APO458378691 Apollo Scientific Ltd

2-Cyclopropylaniline

  • MDL number MFCD00604193
  • Cas Number : 3158-73-4
  • Molecular Weight : 133.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-cyclopropylaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-cyclopropylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart