APO458378692 Apollo Scientific Ltd

3-Cyclopropylaniline

  • MDL number MFCD06802404
  • Cas Number : 485402-64-0
  • Molecular Weight : 133.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-cyclopropylaniline

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/3-cyclopropylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart