APO458955501 Apollo Scientific Ltd

Disodium 3-hydroxynaphthalene-2,7-disulphonate

  • MDL number MFCD00065915
  • Cas Number : 135-51-3
  • Molecular Weight : 348.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name disodium 3-hydroxy-2,7-naphthalenedisulfonate
Inchi Code 1S/C10H8O7S2.2Na/c11-9-4-6-1-2-8(18(12,13)14)3-7(6)5-10(9)19(15,16)17;;/h1-5,11H,(H,12,13,14)(H,15,16,17);;/q;2*+1/p-2
Inchi Key VNEBWJSWMVTSHK-UHFFFAOYSA-L

Description

Synonym 3-Hydroxynaphthalene-2,7-disulphonic acid, disodium salt
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/disodium-3-hydroxynaphthalene-27-disulphonate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/disodium-3-hydroxynaphthalene-27-disulphonate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart