APO458955536 Apollo Scientific Ltd

1-Ethyl-2-iodobenzene

  • MDL number MFCD00040871
  • Cas Number : 18282-40-1
  • Molecular Weight : 232.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.618
Flash Point 101
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 221
Product Link https://store.apolloscientific.co.uk/product/1-ethyl-2-iodobenzene

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-ethyl-2-iodobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart