APO458955563 Apollo Scientific Ltd

2,6-Dimethoxypyrazine

  • MDL number MFCD00955843
  • Cas Number : 4774-15-6
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 48-49
Iupac Name 2,6-dimethoxypyrazine
Inchi Code 1S/C6H8N2O2/c1-9-5-3-7-4-6(8-5)10-2/h3-4H,1-2H3
Inchi Key FCLZSYLOQPRJMS-UHFFFAOYSA-N

Description

Flash Point 75/10mm
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 48-49
Product Link https://store.apolloscientific.co.uk/product/26-dimethoxypyrazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-dimethoxypyrazine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart