APO458955597 Apollo Scientific Ltd

3-Bromo-5-methoxybenzyl alcohol

  • MDL number MFCD00229988
  • Cas Number : 262450-64-6
  • Molecular Weight : 217.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-46
Synonym (3-Bromo-5-methoxyphenyl)methanol, 3-Bromo-5-(hydroxymethyl)anisole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-methoxybenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-methoxybenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart