APO458955618 Apollo Scientific Ltd

3-(Bromomethyl)phenyl acetate

  • MDL number MFCD08543447
  • Cas Number : 49617-80-3
  • Molecular Weight : 229.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at -20
Physical Form Liquid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromomethylphenyl-acetate

Safety Information

signalword Danger
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromomethylphenyl-acetate
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart