APO458955619 Apollo Scientific Ltd

11-(Perfluorooctyl)undecan-1-ol

  • MDL number MFCD28963749
  • Cas Number : 1512-02-3
  • Molecular Weight : 590.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-78
Synonym 12,12,13,13,14,14,15,15,16,16,17,17,18,18,19,19,19-Heptadecafluorononadecan-1-ol, 11-(Heptadecafluorooctyl)undecan-1-ol
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/11-perfluorooctylundecan-1-ol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/11-perfluorooctylundecan-1-ol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart