APO458955620 Apollo Scientific Ltd

1,3-Oxazole-2-carboxaldehyde

  • MDL number MFCD03412587
  • Cas Number : 65373-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43200

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43200
pictograms GHS02;GHS07
hstatements H226;H319
Availability details for
Quantity
  • Availability
  • Add to Cart