APO458955634 Apollo Scientific Ltd

3-(Methylsulphonyl)phenylacetic acid

  • MDL number MFCD03426066
  • Cas Number : 1877-64-1
  • Molecular Weight : 214.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-98
Synonym 3-(Carboxymethyl)phenyl methyl sulphone, [3-(Methylsulphonyl)phenyl]acetic acid
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methylsulphonylphenylacetic-acid

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-methylsulphonylphenylacetic-acid
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart