APO458955641 Apollo Scientific Ltd

1-Methylazetidine-3-carboxylic acid

  • MDL number MFCD11520621
  • Cas Number : 875629-26-8
  • Molecular Weight : 115.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Carboxy-1-methylazetidine, 3-Carboxy-1-methylazetane
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-methylazetidine-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-methylazetidine-3-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart