APO458955704 Apollo Scientific Ltd

4-(Difluoromethoxy)-3-fluorobromobenzene

  • MDL number MFCD11036067
  • Cas Number : 147992-27-6
  • Molecular Weight : 241.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-alpha,alpha,2-trifluoroanisole
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-difluoromethoxy-3-fluorobromobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-difluoromethoxy-3-fluorobromobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart