APO458955709 Apollo Scientific Ltd

4'-(Trifluoroacetyl)acetophenone

  • MDL number MFCD12547944
  • Cas Number : 86988-49-0
  • Molecular Weight : 216.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(4-Acetylphenyl)-2,2,2-trifluoroethan-1-one
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-trifluoroacetylacetophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoroacetylacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart