APO458955717 Apollo Scientific Ltd

4-(Difluoromethoxy)-3-fluorobenzaldehyde

  • MDL number MFCD11847153
  • Cas Number : 1214379-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-difluoromethoxy-3-fluorobenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-difluoromethoxy-3-fluorobenzaldehyde
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart