APO458955720 Apollo Scientific Ltd

4,4,5,5-Tetramethyl-2-(2,3,4-trifluorophenyl)-1,3,2-dioxaborolane

  • MDL number MFCD12405349
  • Cas Number : 881401-96-3
  • Molecular Weight : 258.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4455-tetramethyl-2-234-trifluorophenyl-132-dioxaborolane

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4455-tetramethyl-2-234-trifluorophenyl-132-dioxaborolane
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart