APO458955722 Apollo Scientific Ltd

4,4,5,5-Tetramethyl-2-(2,4,6-trifluorophenyl)-1,3,2-dioxaborolane

  • MDL number MFCD12405351
  • Cas Number : 325143-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-46
Synonym 2,4,6-Trifluorophenylboronic acid, pinacol ester
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=73032

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=73032
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart