APO458955723 Apollo Scientific Ltd

2-(5-Bromo-2-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD12405354
  • Cas Number : 1111096-03-7
  • Molecular Weight : 314.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-71
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-5-bromo-2-fluoro-4-methylphenyl-4455-tetramethyl-132-dioxaborolane

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/2-5-bromo-2-fluoro-4-methylphenyl-4455-tetramethyl-132-dioxaborolane
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart