APO458955727 Apollo Scientific Ltd

3-Bromo-5-methoxybenzyl bromide

  • MDL number MFCD12761608
  • Cas Number : 59297-29-9
  • Molecular Weight : 279.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-5-(bromomethyl)anisole, 1-Bromo-3-(bromomethyl)-5-methoxybenzene, 3-Bromo-5-(bromomethyl)phenyl methyl ether
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-methoxybenzyl-bromide

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-methoxybenzyl-bromide
pictograms GHS05;GHS07
hstatements H302;H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart