APO458955753 Apollo Scientific Ltd

8-Bromo-5-(methylthio)[1,2,4]triazolo[4,3-c]pyrimidine

  • MDL number MFCD28502491
  • Cas Number : 99451-55-5
  • Molecular Weight : 245.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Bromo-5-(methylsulfanyl)[1,2,4]triazolo[4,3-c]pyrimidine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-bromo-5-methylthio124triazolo43-cpyrimidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/8-bromo-5-methylthio124triazolo43-cpyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart