APO459179341 Apollo Scientific Ltd

3-Bromo-4-iodoaniline

  • MDL number MFCD09753717
  • Cas Number : 860435-38-7
  • Molecular Weight : 297.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-iodoaniline

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-iodoaniline
pictograms GHS06
hstatements H301;H312+H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart