APO459179379 Apollo Scientific Ltd

4-Bromo-3-(difluoromethoxy)nitrobenzene

  • MDL number MFCD15526849
  • Cas Number : 835633-47-1
  • Molecular Weight : 268.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-difluoromethoxynitrobenzene

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-difluoromethoxynitrobenzene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart