APO459179392 Apollo Scientific Ltd

2-Bromo-3-chloro-6-fluorobenzaldehyde

  • MDL number MFCD11520667
  • Cas Number : 1056264-66-4
  • Molecular Weight : 237.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-chloro-6-fluorobenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-chloro-6-fluorobenzaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart