APO459179629 Apollo Scientific Ltd

(2-Methoxy-5-phenyl)phenyl isothiocyanate

  • MDL number MFCD00041078
  • Cas Number : 206761-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-81
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=75438

Safety Information

signalword Danger
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=75438
pictograms GHS05;GHS07
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart