APO459179666 Apollo Scientific Ltd

alpha-chlorobenzaldoxime

  • MDL number MFCD02183500
  • Cas Number : 698-16-8
  • Molecular Weight : 155.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-52
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/alpha-chlorobenzaldoxime

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/alpha-chlorobenzaldoxime
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart