APO459179988 Apollo Scientific Ltd

(3,4-Dihydro-1H-isochromen-1-ylmethyl)amine

  • MDL number MFCD00457645
  • Cas Number : 19158-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=75802

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=75802
pictograms GHS05;GHS07
hstatements H302+H312+H332;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart