APO459180275 Apollo Scientific Ltd

4-(2-Methylpyridin-3-yloxy)-6-chloro-5-Methylpyrimidine

  • MDL number MFCD12756519
  • Cas Number : 930093-72-4
  • Molecular Weight : 235.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-5-methyl-6-[(2-methyl-3-pyridinyl)oxy]pyrimidine
Inchi Code 1S/C11H10ClN3O/c1-7-10(12)14-6-15-11(7)16-9-4-3-5-13-8(9)2/h3-6H,1-2H3
Inchi Key SLKKQWWYTQOOHM-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-2-methylpyridin-3-yloxy-6-chloro-5-methylpyrimidine

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS https://store.apolloscientific.co.uk/product/4-2-methylpyridin-3-yloxy-6-chloro-5-methylpyrimidine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart