APO459180847 Apollo Scientific Ltd

Ethyl chloro[(4-fluorophenyl)hydrazono]acetate

  • MDL number MFCD06809227
  • Cas Number : 37522-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=76657

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=76657
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart