APO459181860 Apollo Scientific Ltd

3-Cyanobicyclo[1.1.1]pentane-1-carboxylic acid

  • MDL number MFCD19229172
  • Cas Number : 83249-02-9
  • Molecular Weight : 137.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-cyanobicyclo[1.1.1]pentane-1-carboxylic acid
Inchi Code 1S/C7H7NO2/c8-4-6-1-7(2-6,3-6)5(9)10/h1-3H2,(H,9,10)
Inchi Key MFBJJYLOKZSNPI-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-cyanobicyclo111pentane-1-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-cyanobicyclo111pentane-1-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart