APO459181861 Apollo Scientific Ltd

1-(Bromomethyl)bicyclo[1.1.1]pentane

  • MDL number MFCD20633000
  • Cas Number : 161043-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=77641

Safety Information

signalword Warning
pcodes P280;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=77641
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart