APO459181881 Apollo Scientific Ltd

2-(Bicyclo[1.1.1]pentan-1-yl)acetic acid

  • MDL number MFCD19229645
  • Cas Number : 131515-31-6
  • Molecular Weight : 126.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bicyclo111pentan-1-ylacetic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bicyclo111pentan-1-ylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart