APO459379816 Apollo Scientific Ltd

3,4-Dimethoxyphenacyl bromide

  • MDL number MFCD03425186
  • Cas Number : 1835-02-5
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-1-(3,4-dimethoxyphenyl)ethanone
Inchi Code 1S/C10H11BrO3/c1-13-9-4-3-7(8(12)6-11)5-10(9)14-2/h3-5H,6H2,1-2H3
Inchi Key NUAIPKMBWNVQIM-UHFFFAOYSA-N

Description

Synonym 2-Bromo-1-(3,4-dimethoxyphenyl)ethan-1-one
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/34-dimethoxyphenacyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/34-dimethoxyphenacyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart