APO459379828 Apollo Scientific Ltd

Oxan-3-amine

  • MDL number MFCD07371531
  • Cas Number : 120811-32-7
  • Molecular Weight : 101.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tetrahydro-2H-pyran-3-ylamine
Inchi Code 1S/C5H11NO/c6-5-2-1-3-7-4-5/h5H,1-4,6H2
Inchi Key WUUOEJJGRCQGBQ-UHFFFAOYSA-N

Description

Synonym Tetrahydro-2H-pyran-3-amine
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/oxan-3-amine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/oxan-3-amine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart