APO459379849 Apollo Scientific Ltd

1,1,5,5-Tetrafluoropentane-2,4-dione

  • MDL number MFCD03419795
  • Cas Number : 70086-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,1-Difluoro-3-(difluoroacetyl)acetone, 2,4-Dioxo-1,1,5,5-tetrafluoropentane, 1,1,5,5-Tetrafluoroacetylacetone
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51478

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51478
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart