APO465886736 Apollo Scientific Ltd

5-Bromo-2,4-difluorostyrene

  • MDL number MFCD29049257
  • Cas Number : 1935371-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=78089

Safety Information

signalword Warning
pcodes P280;P302+P352;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=78089
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart