APO465886745 Apollo Scientific Ltd

2-Amino-4-bromo-5-fluorobenzonitrile

  • MDL number MFCD29049266
  • Cas Number : 1935427-48-7
  • Molecular Weight : 215.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-bromo-5-fluorobenzonitrile

Safety Information

signalword Warning
pcodes P260;P280;P301+P312;P302+P352;P304+P340;P361+P364
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-bromo-5-fluorobenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart