APO465886749 Apollo Scientific Ltd

2-Bromo-3,5-difluorobenzyl bromide

  • MDL number MFCD28737918
  • Cas Number : 1807071-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=78119

Safety Information

signalword Danger
pcodes P301+P330+P331;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=78119
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart