APO465886932 Apollo Scientific Ltd

1,1,1-Trifluoro-3-(1H-pyrazol-1-yl)propan-2-ol

  • Cas Number : 1342051-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=77977

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=77977
Availability details for
Quantity
  • Availability
  • Add to Cart