APO465886933 Apollo Scientific Ltd

1,1,1-Trifluoro-3-[4-(2-hydroxyethyl)piperazin-1-yl]propan-2-ol

  • Cas Number : 477847-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=77978

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=77978
Availability details for
Quantity
  • Availability
  • Add to Cart