APO465886964 Apollo Scientific Ltd

3-(Difluoromethoxy)-4-fluoronitrobenzene

  • MDL number MFCD14698450
  • Cas Number : 97963-50-3
  • Molecular Weight : 207.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name difluoromethyl 2-fluoro-5-nitrophenyl ether
Inchi Code 1S/C7H4F3NO3/c8-5-2-1-4(11(12)13)3-6(5)14-7(9)10/h1-3,7H
Inchi Key UKSAGCUNNXEOBJ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-difluoromethoxy-4-fluoronitrobenzene

Safety Information

signalword Warning
pcodes P280;P301+P312;P261
MSDS https://store.apolloscientific.co.uk/product/3-difluoromethoxy-4-fluoronitrobenzene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart