APO465886969 Apollo Scientific Ltd

2-(Difluoromethoxy)-3-fluoroaniline

  • MDL number MFCD14698441
  • Cas Number : 153468-97-4
  • Molecular Weight : 177.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-difluoromethoxy-3-fluoroaniline

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-difluoromethoxy-3-fluoroaniline
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart