APO465886989 Apollo Scientific Ltd

(R)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole 1,1-dioxide

  • MDL number MFCD26406787
  • Cas Number : 143577-46-2
  • Molecular Weight : 163.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/r-456-tetrahydro-3h-pyrrolo12-coxathiazole-11-dioxide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/r-456-tetrahydro-3h-pyrrolo12-coxathiazole-11-dioxide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart