APO465887014 Apollo Scientific Ltd

2-Bromo-3,6-difluorotoluene

  • MDL number MFCD12922624
  • Cas Number : 1208074-75-2
  • Molecular Weight : 207.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-36-difluorotoluene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-36-difluorotoluene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart