APO465887030 Apollo Scientific Ltd

2-Bromo-3,4-difluorobenzaldehyde

  • MDL number MFCD28669844
  • Cas Number : 1578156-21-4
  • Molecular Weight : 221

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-34-difluorobenzaldehyde

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS https://store.apolloscientific.co.uk/product/2-bromo-34-difluorobenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart