APO465887040 Apollo Scientific Ltd

2-Bromo-3,5-difluorobenzoic acid

  • MDL number MFCD09835211
  • Cas Number : 651027-01-9
  • Molecular Weight : 237

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-3,5-difluorobenzoic acid
Inchi Code 1S/C7H3BrF2O2/c8-6-4(7(11)12)1-3(9)2-5(6)10/h1-2H,(H,11,12)
Inchi Key KBOFTBQUYXDFEF-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-35-difluorobenzoic-acid

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/2-bromo-35-difluorobenzoic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart