APO465887103 Apollo Scientific Ltd

2-Bromo-4-fluoro-6-iodoaniline

  • MDL number MFCD24160442
  • Cas Number : 1447606-71-4
  • Molecular Weight : 315.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-fluoro-6-iodoaniline

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-fluoro-6-iodoaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart