APO465887104 Apollo Scientific Ltd

2-Bromo-4-fluoro-6-methoxyaniline

  • MDL number MFCD27949835
  • Cas Number : 354574-32-6
  • Molecular Weight : 220.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-fluoro-6-methoxyaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-fluoro-6-methoxyaniline
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart