APO466131503 Apollo Scientific Ltd

3-Methoxy-2-methylbenzoic acid

  • MDL number MFCD02094039
  • Cas Number : 55289-06-0
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 149-151
Iupac Name 3-methoxy-2-methylbenzoic acid
Inchi Code 1S/C9H10O3/c1-6-7(9(10)11)4-3-5-8(6)12-2/h3-5H,1-2H3,(H,10,11)
Inchi Key JPCISVSOTKMFPG-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 149-151
Product Link https://store.apolloscientific.co.uk/product/3-methoxy-2-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-methoxy-2-methylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart