APO466131509 Apollo Scientific Ltd

4-(Aminomethyl)-1-methylpiperidine dihydrochloride

  • MDL number MFCD11035762
  • Cas Number : 1187582-53-1
  • Molecular Weight : 201.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1-methyl-4-piperidinyl)methanamine dihydrochloride
Inchi Code 1S/C7H16N2.2ClH/c1-9-4-2-7(6-8)3-5-9;;/h7H,2-6,8H2,1H3;2*1H
Inchi Key QJOPPLGHTCLBJO-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-aminomethyl-1-methylpiperidine-dihydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-aminomethyl-1-methylpiperidine-dihydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart