APO466131513 Apollo Scientific Ltd

5-Bromo-2,4-difluorobenzaldehyde

  • MDL number MFCD09842439
  • Cas Number : 473416-91-0
  • Molecular Weight : 221

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2,4-difluorobenzaldehyde
Inchi Code 1S/C7H3BrF2O/c8-5-1-4(3-11)6(9)2-7(5)10/h1-3H
Inchi Key UFNBTKFAPDUBCY-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-24-difluorobenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-24-difluorobenzaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart