APO466131521 Apollo Scientific Ltd

Potassium (3-acetylphenyl)trifluoroborate

  • MDL number MFCD07368352
  • Cas Number : 854906-74-4
  • Molecular Weight : 226.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/potassium-3-acetylphenyltrifluoroborate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/potassium-3-acetylphenyltrifluoroborate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart